Learn More
Beperidium iodide, MedChemExpress
MedChemExpress Beperidium iodide shows a competitive antagonistic effect against acetylcholine receptor with a pA2 of 7.93.
Brand:  Medchem Express HY-100152
Encompass_Preferred
 View more versions of this product
Product Code. 16422808
  • £751.41 / 10mg
Explore more special offers
Description
Description
Beperidium iodide shows a competitive antagonistic effect against acetylcholine receptor with a pA2 of 7.93.
Specifications
Specifications
Beperidium iodide
White
C23H34IN3O3
SX 810
C[N+]1(CC)CCC(OC(C(C2=NOC3=C2C=CC=C3)N4CCCCCC4)=O)CC1.[I-]
527.44
Research
COVID-19-immunoregulation
86434-57-3
Research
10 mg
DMSO : ≥ 100 mg/mL (189.60 mM)
527.44
99.79%
Solid
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Beperidium iodide, MedChemExpress