missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ FetalClone™ III Serum (U.S.)
Designed for fibroblast applications, supplement cell culture growth with U.S.-sourced HyClone™ FetalClone™ III Serum, the most widely applicable fetal bovine serum (FBS) alternative.
Brand: Cytiva SH30109.02
94.53 GBP valid until 2024-12-31
Use promo code "21631" to get your promotional price.
This item is not returnable.
View return policy
Description
- Economical alternative to FBS
- Performance comparable with FBS
- High lot-to-lot consistency
- Low endotoxin and hemoglobin levels
Specifications
Albumin, Alkaline Phosphatase, Blood Urea Nitrogen, Calcium, Chloride, Creatinine, Gamma Globulin, Glucose, IgG, Inorganic Phosphorus, Iron, Lactate Dehydrogenase, Osmolality, Percent Iron Saturation, pH, Potassium, SGOT, SGPT, Sodium, TIBC, Total Bilirubin, Total Protein | |
Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2 to 8°C. | |
FBS Replacement Serum | |
Sterile | |
None | |
≤ 10 EU/mL | |
≤ 20 mg/dL | |
HyClone™ | |
USDA Inspected |
Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities | |
United States | |
100 mL | |
Sequential-filtered, 100 nm | |
Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus | |
No | |
Typically less than 2 μg/mL | |
No |
Safety and Handling
Hazard Statement
Supplemental information
- EUH210-Safety data sheet available on request.
Supplemental information
- UNKNOWN ACUTE-9.9 per cent of the mixture consists of component(s) of unknown acute toxicity.
- UNKNOWN AQUATIC-Contains 9.9% of components with unknown hazards to the aquatic environment.