Learn More
Berkshire™ SatPax™ 3000 Presaturated Specialty Wipers
For use in ISO Class 3 to 5 and higher cleanroom environments
Brand: Berkshire™ SPX3000.001.12
31.90 GBP valid until 2024-07-31
Use promo code "21640" to get your promotional price.
Code : 04
Additional Details : CAS Number : 67-63-0 Weight : 0.15000kg Transport : UN number : 3175 Chem class : 4.1 Pack group : II
Description
- Knitted polyester wipers resolve safety issues associated with storing, mixing and dispensing solvents; also improve the cleaning process
- Patented sealed edge process for reduced fiber contamination
- Presaturated with consistent IPA/DI Water concentrations and saturation
Order Info
Measure 9 x 9 (23 x 23cm). Available 30 J-folded sheets per resealable flexible pouch.
Specifications
Pre-saturated IPA Wipers | |
22.86 cm | |
22.86 cm | |
145 | |
Sealed | |
For use in ISO Class 3 to 5 and higher cleanroom environments, for use in wet cleaning of critical surfaces where control of flammable solvents and flammable solvent concentrations is required | |
70% Isopropyl Alcohol, 30% Deionised Water |
9 in. | |
9 in. | |
Polyester Knit | |
RoHS, REACH | |
9 x 9 in. (22.86 x 22.86 cm) | |
2-ply |
Safety and Handling
- SatPax 70% IPA / 30% DI H2O
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable solid Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H228-Flammable solid.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
Precautionary Statement
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : isopropyl alcohol