missing translation for 'onlineSavingsMsg'
Learn More
beta-Glycerophosphate, 200mM soln., Thermo Scientific Chemicals Product Code.: 15444559

beta-Glycerophosphate, 200mM soln., Thermo Scientific Chemicals

Product Code. 15444559
100 mL
Click to view available options
Quantity:
50 mL
100 mL
This item is not returnable. View return policy

Product Code. 15444559

Brand: Thermo Scientific Alfa Aesar J62121.AE

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

β-Glycerolphosphate (Glycerol-2-Phosphate) is used in the development of hydrogels and scaffolds that have applications in tissue engineering and cell growth and differentiation. It has also been used as an additive in isolation mediums, similarly providing available phosphate to the isolate. It is a phosphate group donor in matrix mineralization studies that acts as a protein phosphatase inhibitor. Accelerates calcification in vascular smooth muscle cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
β-Glycerolphosphate (Glycerol-2-Phosphate) is used in the development of hydrogels and scaffolds that have applications in tissue engineering and cell growth and differentiation. It has also been used as an additive in isolation mediums, similarly providing available phosphate to the isolate. It is a phosphate group donor in matrix mineralization studies that acts as a protein phosphatase inhibitor. Accelerates calcification in vascular smooth muscle cells.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 819-83-0
Molecular Formula C3H9Na2O6P
Molecular Weight (g/mol) 218.052
MDL Number MFCD00002135
InChI Key UQVSFONYLAZJDB-UHFFFAOYSA-N
Synonym glycerol 2-phosphate disodium salt hydrate, disodium glycerol 2-phosphate hydrate, beta-glycerophosphoric acid disodium salt, c3h7o6p.2na.h2o, sodium glycerophosphate usan, b-glycerophosphate disodium salt hydrate, sodium 1,3-dihydroxypropan-2-yl phosphate hydrate 2:1:x, beta-glycerophosphate disodium salt hydrate, ≤1.0 mol % l-alpha-isomer, beta-glycerophosphate disodium salt hydrate, powder, bioreagent, suitable for cell culture, beta-glycerophosphate disodium salt hydrate, ≤2.4 mol % alpha-isomer based on c3h7o6pna2 with x h2oShow MoreShow Less
PubChem CID 131860121
IUPAC Name 1,3-dihydroxypropan-2-yl dihydrogen phosphate;sodium
SMILES C(C(CO)OP(=O)(O)O)O.[Na].[Na]

Specifications

Color Colorless
Quantity 100 mL
Solubility Information Soluble in water.
Formula Weight 216.04
Concentration or Composition (by Analyte or Components) 200mM soln.
Physical Form Liquid
Chemical Name or Material beta-Glycerophosphate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZ27K527 Certificate TypeCertificate of Analysis Date23/01/2025 Catalog NumberJ62121.AD,J62121.AE
Lot NumberX23K533 Certificate TypeCertificate of Analysis Date30/10/2024 Catalog NumberJ62121.AD,J62121.AE
Lot NumberW21K529 Certificate TypeCertificate of Analysis Date20/09/2024 Catalog NumberJ62121.AD,J62121.AE
Lot NumberR09K533 Certificate TypeCertificate of Analysis Date14/06/2024 Catalog NumberJ62121.AD,J62121.AE
Lot NumberN25K546 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog NumberJ62121.AD,J62121.AE

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
beta-Glycerophosphate, 200mM soln., Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.