missing translation for 'onlineSavingsMsg'
Learn More
beta-Sitosterol, with ca. 10% campesterol, ca. 75% beta-sitosterol, Thermo Scientific Chemicals Product Code.: 11964641

beta-Sitosterol, with ca. 10% campesterol, ca. 75% beta-sitosterol, Thermo Scientific Chemicals

Product Code. 11964641
500 g, Plastic bottle
Click to view available options
Quantity:
500 g
Packaging:
Plastic bottle
This item is not returnable. View return policy

Product Code. 11964641

Brand: Thermo Scientific Acros 132725000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 83-46-5
Color White
Loss on Drying 3.5% max.
Infrared Spectrum Authentic
Packaging Plastic bottle
Molecular Formula C29H50O
Quantity 500 g
Specific Rotation Condition − 28.00 (20.00°C c=2,CHCl3)
Specific Rotation -24° to -32° (20°C, 589nm) (c=2, CHCl3) Typically
Synonym beta-sitosterol, sitosterol, cupreol, quebrachol, 22,23-dihydrostigmasterol, azuprostat, triastonal, cinchol, rhamnol, harzol
InChI Key KZJWDPNRJALLNS-VJSFXXLFSA-N
SMILES CCC(CCC(C)C1CCC2C1(CCC3C2CC=C4C3(CCC(C4)O)C)C)C(C)C
IUPAC Name (3S,8S,9S,10R,13R,14S,17R)-17-[(2R,5R)-5-ethyl-6-methylheptan-2-yl]-10,13-dimethyl-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-ol
Molecular Weight (g/mol) 414.718
PubChem CID 222284
ChEBI CHEBI:27693
Formula Weight 414.7
Percent Purity ≥75%
Physical Form Powder
Chemical Name or Material β-Sitosterol
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0450466 Certificate TypeCertificate of Analysis Date27/02/2024 Catalog Number132721000,132725000,132720050
Lot NumberA0421663 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number132721000,132725000,132720050
Lot NumberA0403942 Certificate TypeCertificate of Analysis Date07/01/2021 Catalog Number132721000,132725000,132720050
Lot NumberA0366899 Certificate TypeCertificate of Analysis Date10/06/2020 Catalog Number132721000,132725000,132720050

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
beta-Sitosterol, with ca. 10% campesterol, ca. 75% beta-sitosterol, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.