missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ BetaBasic™ 18 Guard Cartridges

Available in limited dimensions. Please contact Customer Service for more details.

Brand:  Thermo Scientific™ 71505-013001

Code : 16

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10438094

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

Thermo Scientific™ BetaBasic™ 18 Guard Cartridges are available in limited dimensions. Please contact Customer Service for more details.

  • The 10mm design offers maximum protection with minimal increase in retention
  • Fit Thermo Scientific UNIGUARD™ direct connection and stand alone holders
  • Highly efficient and reproducible Thermo Scientific columns
  • 150Å pore size for small molecules, peptides and protein digests
  • Superb pH stability

Requires:

Thermo Scientific Unigard™ direct connection or stand-alone holder, sold separately.

Specifications

Specifications

13%
5 μm
150Å
3 mm
Yes
2 to 12
10 mm
200 m2/g
Uniguard Direct-connection and Stand Alone Holder
L1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers