Learn More
Thermo Scientific™ BetaBasic™ 18 Guard Cartridges
Available in limited dimensions. Please contact Customer Service for more details.
£229.62 - £393.20
Specifications
Carbon Load | 13% |
---|---|
pH | 2 to 12 |
Length (Metric) | 10 mm |
Pore Size | 150Å |
Surface Area | 200 m2/g |
Product Code | Brand | Diameter (Metric) | Length (Metric) | Particle Size | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Length (Metric) | Particle Size | Price | Quantity & Availability | |||
10197984
|
Thermo Scientific™
71503-012101 |
2.1 mm | 10 mm | 3 μm |
£393.20
Pack of 4 |
Estimated Shipment: 01-08-2024
Log in to see stock. |
|||
11582841
|
Thermo Scientific™
71503-014001 |
4 mm | 10 mm | 3 μm |
£393.20
Pack of 4 |
Estimated Shipment: 01-08-2024
Log in to see stock. |
|||
11592841
|
Thermo Scientific™
71505-014001 |
4 mm | 10 mm | 5 μm |
£353.77
Pack of 4 |
In Stock
Log in to see stock. |
|||
10419634
|
Thermo Scientific™
71505-012101 |
2.1 mm | 10 mm | 5 μm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
10438094
|
Thermo Scientific™
71505-013001 |
3 mm | 10 mm | 5 μm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
11572841
|
Thermo Scientific™
71503-013001 |
3 mm | 10 mm | 3 μm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
Thermo Scientific™ BetaBasic™ 18 Guard Cartridges are available in limited dimensions. Please contact Customer Service for more details.
- The 10mm design offers maximum protection with minimal increase in retention
- Fit Thermo Scientific UNIGUARD™ direct connection and stand alone holders
- Highly efficient and reproducible Thermo Scientific columns
- 150Å pore size for small molecules, peptides and protein digests
- Superb pH stability
Requires:
Thermo Scientific Unigard™ direct connection or stand-alone holder, sold separately.
Specifications
13% | |
10 mm | |
200 m2/g | |
Yes |
2 to 12 | |
150Å | |
Uniguard Direct-connection and Stand Alone Holder | |
L1 |