missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ BetaBasic™ CN Particle Size Guard Cartridges, 5μm Particle Size

Thermo Scientific™ BetaBasic™ CN Particle Size Guard Cartridges are available in limited dimensions. Please contact Customer Service for more details.

Brand:  Thermo Scientific™ 71705-014001

 View more versions of this product

Product Code. 10321115

  • £382.91 / Pack of 4
Estimated Shipment: 24-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Thermo Scientific™ BetaBasic™ CN Particle Size Guard Cartridges are available in limited dimensions. Please contact Customer Service for more details.

  • The 10mm design offers maximum protection with minimal increase in retention
  • Fit Thermo Scientific UNIGUARD™ direct connection and stand alone holders
  • Highly efficient and reproducible Thermo Scientific columns
  • 150Å pore size for small molecules, peptides and protein digests
  • Superb pH stability
TRUSTED_SUSTAINABILITY
Specifications

Specifications

5%
5 μm
150Å
4 mm
Yes
Beta
2 to 9
10 mm
200 m2/g
Uniguard Direct-connection and Stand Alone Holder
L10
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers