missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ BetaBasic™ CN Particle Size Guard Cartridges, 5μm Particle Size
Thermo Scientific™ BetaBasic™ CN Particle Size Guard Cartridges are available in limited dimensions. Please contact Customer Service for more details.
£362.97
Specifications
Carbon Load | 5% |
---|---|
pH | 2 to 9 |
Particle Size | 5 μm |
Length (Metric) | 10 mm |
Pore Size | 150Å |
Product Code | Brand | Diameter (Metric) | Length (Metric) | Particle Size | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Length (Metric) | Particle Size | Price | Quantity & Availability | |||
10321115
|
Thermo Scientific™
71705-014001 |
4 mm | 10 mm | 5 μm |
£362.97
Pack of 4 |
Estimated Shipment: 21-08-2024
Log in to see stock. |
|||
Description
Thermo Scientific™ BetaBasic™ CN Particle Size Guard Cartridges are available in limited dimensions. Please contact Customer Service for more details.
- The 10mm design offers maximum protection with minimal increase in retention
- Fit Thermo Scientific UNIGUARD™ direct connection and stand alone holders
- Highly efficient and reproducible Thermo Scientific columns
- 150Å pore size for small molecules, peptides and protein digests
- Superb pH stability
Specifications
5% | |
5 μm | |
150Å | |
Uniguard Direct-connection and Stand Alone Holder | |
L10 |
2 to 9 | |
10 mm | |
200 m2/g | |
Yes |