missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ BetaSil™ C8 Guard Cartridges

Drop-in guard cartridges offer convenience, economy and effective protection for extending column lifetime.

Brand:  Thermo Scientific™ 70205-014001

 View more versions of this product

Product Code. 10448864

  • £379.89 / Pack of 4
Estimated Shipment: 11-12-2024
to see stock.
Explore more special offers
Add to basket

Requires : Thermo Scientific UNIGUARD™ direct connection or stand alone holder, sold separately.
This item is not returnable. View return policy

Description

Description

Drop-in guard cartridges offer convenience, economy and effective protection for extending column lifetime.

  • The 10mm design offers maximum protection with minimal increase in retentio
  • Fit Thermo Scientific UNIGUARD™ direct connection and stand alone holders
  • Particle size: 5μm
  • 10L x 3.0mm I.D.
  • Thermo Scientific chemistry on 100Å silica
  • High surface area with high bonded phase coverage

TRUSTED_SUSTAINABILITY
Specifications

Specifications

5 μm
4 or 4.6 mm
BetaSil
10 mm
C8
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers