Learn More
Buerkle™ Power Supply Unit for Buerkle™ AccuOne/EnergyOne
Buerkle™ Power Supply Unit for Buerkle™ AccuOne/EnergyOne that includes an international plug adapter.
Brand:  Buerkle™ 5900-3020
Product Code. 11961641
  • £195.00 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Power Supply Unit for Buerkle™ AccuOne/EnergyOne
  • Power supply via 230V power supply unit
  • International plug adapter included with delivery
Specifications
Specifications
Power Supply Unit
230 V
0.01 kg
Accu/EnergyOne Barrel Pump
Power supply unit incl. Connectors for Accu/EnergyOne
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Buerkle™ Power Supply Unit for Buerkle™ AccuOne/EnergyOne > Voltage: 230V