missing translation for 'onlineSavingsMsg'
Learn More

Bicinchoninic acid disodium salt, Thermo Scientific Chemicals

For determination of Cu and protein assays

£88.94 - £346.73

Chemical Identifiers

CAS 979-88-4
Molecular Formula C20H10N2Na2O4
Molecular Weight (g/mol) 388.29
MDL Number MFCD00037500
InChI Key AUPXFICLXPLHBB-UHFFFAOYSA-L
Synonym sodium 2,2'-biquinoline-4,4'-dicarboxylate, 2,2'-biquinoline-4,4-dicarboxylic acid disodium salt, 2,2'-biquinoline-4,4'-dicarboxylic acid, disodium salt, sodium bicinchoninate, 2,2'-biquinoline-4,4'-dicarboxylic acid disodium salt, disodium 2,2'-biquinoline-4,4'-dicarboxylate, bicinchoninic acid disodium salt, bca, 2,2'-biquinoline-4,4'-dicarboxylic acid, sodium salt 1:2
PubChem CID 164763
SMILES [Na+].[Na+].[O-]C(=O)C1=C2C=CC=CC2=NC(=C1)C1=CC(C([O-])=O)=C2C=CC=CC2=N1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11359778
View Documents Promotion Details
Thermo Scientific Alfa Aesar
044247.04
2 g
£88.94
2g
Please to purchase this item. Need a web account? Register with us today!
11369778
View Documents
Thermo Scientific Alfa Aesar
044247.09
10 g
£346.73
10g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

For determination of Cu and protein assays.Bicinchoninic acid disodium salt is useful for the determination of copper. It is also used for the determination of protein concentration.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

979-88-4
388.29
AUPXFICLXPLHBB-UHFFFAOYSA-L
164763
C20H10N2Na2O4
MFCD00037500
sodium 2,2'-biquinoline-4,4'-dicarboxylate, 2,2'-biquinoline-4,4-dicarboxylic acid disodium salt, 2,2'-biquinoline-4,4'-dicarboxylic acid, disodium salt, sodium bicinchoninate, 2,2'-biquinoline-4,4'-dicarboxylic acid disodium salt, disodium 2,2'-biquinoline-4,4'-dicarboxylate, bicinchoninic acid disodium salt, bca, 2,2'-biquinoline-4,4'-dicarboxylic acid, sodium salt 1:2
[Na+].[Na+].[O-]C(=O)C1=C2C=CC=CC2=NC(=C1)C1=CC(C([O-])=O)=C2C=CC=CC2=N1

Specifications

979-88-4
MFCD00037500
Soluble in water.
[Na+].[Na+].[O-]C(=O)C1=C2C=CC=CC2=NC(=C1)C1=CC(C([O-])=O)=C2C=CC=CC2=N1
388.29
388.29
Bicinchoninic acid disodium salt
C20H10N2Na2O4
sodium 2,2'-biquinoline-4,4'-dicarboxylate, 2,2'-biquinoline-4,4-dicarboxylic acid disodium salt, 2,2'-biquinoline-4,4'-dicarboxylic acid, disodium salt, sodium bicinchoninate, 2,2'-biquinoline-4,4'-dicarboxylic acid disodium salt, disodium 2,2'-biquinoline-4,4'-dicarboxylate, bicinchoninic acid disodium salt, bca, 2,2'-biquinoline-4,4'-dicarboxylic acid, sodium salt 1:2
AUPXFICLXPLHBB-UHFFFAOYSA-L
disodium [2,2'-biquinoline]-4,4'-dicarboxylate
164763
Chunks and/or Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • Bicinchoninic acid, disodium salt

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : Bicinchoninic acid,disodium salt

SDS
Documents

Documents

Certificates

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.