missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals BICINE, 99%

£35.18 - £396.98

Chemical Identifiers

CAS 150-25-4
Molecular Formula C6H12NNaO4
Molecular Weight (g/mol) 185.16
MDL Number MFCD00004295
InChI Key MFBDBXAVPLFMNJ-UHFFFAOYSA-M
PubChem CID 8761
ChEBI CHEBI:40957
IUPAC Name 2-[bis(2-hydroxyethyl)amino]acetic acid
SMILES [Na+].OCCN(CCO)CC([O-])=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11467157
SDS View Documents
Thermo Scientific Chemicals
A14957.18
50 g
£35.18
50g
Please to purchase this item. Need a web account? Register with us today!
11477157
SDS View Documents Promotion Details
Thermo Scientific Chemicals
A14957.30
250 g
£130.66
250g
Please to purchase this item. Need a web account? Register with us today!
11457157
SDS View Documents Promotion Details
Thermo Scientific Chemicals
A14957.0B
1000 g
£396.98
1000g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Bicine is widely utilized as a buffering agent for low temperature work. It is used in kinetic study of a quaternary transition-state analogue complex of creatine kinase reaction. It is employed in the thin layer ion exchange chromatography method for protein resolution. It is employed in serum guanase determination by the preparation of stable substrate solution.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bicine is widely utilized as a buffering agent for low temperature work. It is used in kinetic study of a quaternary transition-state analogue complex of creatine kinase reaction. It is employed in the thin layer ion exchange chromatography method for protein resolution. It is employed in serum guanase determination by the preparation of stable substrate solution.

Solubility
Soluble in water.

Notes
Store in cool place. Incompatible with strong oxidizing agents.
Specifications
Show More
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 205-755-1

missing translation for 'rtecsNumber' : MB9700000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals BICINE, 99%

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.