Learn More
Applied Biosystems™ BigDye™ Terminator v1.1 & v3.1 5X Sequencing Buffer
This sequencing buffer is optimized for use with BigDye™ Terminator v1.1 and v3.1 Cycle Sequencing Kits.
Brand: Applied Biosystems™ 4336697
54.65 GBP valid until 2024-12-31
Use promo code "22915" to get your promotional price.
Additional Details : Weight : 0.00230kg
Description
- Optimized for use with BigDye™ Terminator Cycle Sequencing Kits
Order Info
Each
Specifications
Sequencing Buffer | |
Store at 2°C to 8°C. | |
Standard Sequencing | |
Sequencing Buffer | |
Room Temperature |
5X | |
GeneAmp 9700, Veriti Thermal Cycler, 3500 Genetic Analyzer, 3730xl DNA Analyzer, 3130xl Genetic Analyzer, 3500xL Genetic Analyzer, 3730 DNA Analyzer | |
BigDye™ | |
1 mL | |
Liquid |
For Research Use Only. Not for use in diagnostic procedures.