missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ BigDye™ Terminator v1.1 & v3.1 5X Sequencing Buffer

This sequencing buffer is optimized for use with BigDye™ Terminator v1.1 and v3.1 Cycle Sequencing Kits.

Brand:  Applied Biosystems™ 4336699

Additional Details : Weight : 0.03860kg

 View more versions of this product

Product Code. 15939182

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

  • Optimized for use with BigDye™ Terminator Cycle Sequencing Kits

Order Info

Each

Specifications

Specifications

Sequencing Buffer
Store at 2°C to 8°C.
Standard Sequencing
Sequencing Buffer
Room Temperature
5X
GeneAmp 9700, Veriti Thermal Cycler, 3500 Genetic Analyzer, 3730xl DNA Analyzer, 3130xl Genetic Analyzer, 3500xL Genetic Analyzer, 3730 DNA Analyzer
BigDye™ Terminator, BigDye™
28 mL
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.