missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ BigDye™ Terminator v1.1 & v3.1 5X Sequencing Buffer

This sequencing buffer is optimized for use with BigDye™ Terminator v1.1 and v3.1 Cycle Sequencing Kits.

Brand:  Applied Biosystems™ 4336701

Additional Details : Weight : 0.28580kg

 View more versions of this product

Product Code. 16355292

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

  • Optimized for use with BigDye™ Terminator Cycle Sequencing Kits

Order Info

Each

Specifications

Specifications

Sequencing Buffer
Store at 2°C to 8°C.
Standard Sequencing
233 mL
Liquid
5X
3130xl, 3500, 3500xL Genetic Analyzer; GeneAmp™ 9700;Veriti™ Thermal Cycler; 3730, 3730xl DNA Analyzer
BigDye™ Terminator, BigDye™
Room Temperature
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.