missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ BigDye XTerminator™ Purification Kit

The BigDye XTerminator™ Purification Kit is a fast, simple purification method for DNA sequencing reactions that removes unincorporated BigDye™ terminators and salts.

£219.00 - £17100.00

Specifications

Format Kit
Sample Type Sequencing Products
Final Product Type DNA
For Use With (Application) Sequencing
Includes 2 Bottles
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15741826
View Documents
Applied Biosystems™
4376486
100 Preps
£219.00
2mL
Please to purchase this item. Need a web account? Register with us today!
15899816
View Documents
Applied Biosystems™
4376487
1000 Preps
£1548.00
Each
Please to purchase this item. Need a web account? Register with us today!
15879816
View Documents
Applied Biosystems™
4376484
2500 Preps
£2655.00
Each
Please to purchase this item. Need a web account? Register with us today!
15889816
View Documents
Applied Biosystems™
4376485
40,000 Preps
£17100.00
Each
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Eliminate liquid transfer - Use a single plate
  • Obtain complete dye blob removal
  • Stabilize the samples before analysis
  • Improve your sequencing workflow
  • Perform manual or automated protocols
  • Get rapid, reliable, and reproducible results

Order Info

Each

Specifications

Specifications

Kit
DNA
2 Bottles
Sequencing Reactions
Sequencing Products
Sequencing
BigDye XTerminator™, BigDye™
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Applied Biosystems™ BigDye XTerminator™ Purification Kit

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.