Learn More
Emma Safety Footwear Billy Safety Shoes
All purpose safety boot repels acid and oil like an umbrella repels water. Strong, ankle supporting upper fits the foot snugly and comfortably.
Brand:  Emma Safety Footwear MM73084849
 View more versions of this product
Product Code. 15988951
  • £151.00 / 1 pair
Estimated Shipment: 28-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • S3-Safety shoe with sole plate for wet work
  • Full grain leather construction
  • GripForce™ Duo Heat Resistant Outsole
  • Hydro-Tec™ Sanitized Silver for drier, fresher feet
  • Sanitized
Specifications
Specifications
EN ISO 20345:2011
Black
Hydro-Tec™ Sanitized Silver
49
Laces
Billy
Leather
Steel