Learn More
Thermo Scientific™ BioBasic™ SCX HPLC Columns
BioBasic SCX LC Columns are optimized for the separation of proteins, peptides and other cationic species.
Brand: Thermo Scientific™ 73205-152130
663.10 GBP valid until 2024-10-31
Use promo code "21692" to get your promotional price.
Description
Use Thermo Scientific™ BioBasic™ SCX LC Columns for the optimized separation of proteins, peptides and other cationic species. These high-performance columns are ideal for the analysis of proteins, peptides and other biomolecules with higher efficiency than polymeric ion exchange.
- Strong cation exchange phase based on sulfonic acid chemistry
- Separation and retention of basic and other cationic species
- 300Å pore size for enhanced protein and peptide separations
- Outstanding stability under demanding pH conditions
Ordering Information:
Other column dimensions are available. Please call Customer Service for more information.
Specifications
Analytical Column | |
150 mm | |
5 μm | |
300 Å | |
60°C | |
Sulfonic Acid | |
Spherical, Fully Porous, Ultrapure Silica | |
Ion Exchange | |
Ion Exchange |
2.1 mm | |
5800 psi (400 bar) | |
2 to 8 | |
100 m²/g | |
L52 | |
BioBasic | |
3% | |
No | |
Analytical Column |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.