Learn More
Thermo Scientific™ BioBasic™ SCX HPLC Columns
BioBasic SCX LC Columns are optimized for the separation of proteins, peptides and other cationic species.
£263.16 - £974.85
Specifications
Max. Pressure | 5800 psi (400 bar) |
---|---|
Particle Size | 5 μm |
pH | 2 to 8 |
Pore Size | 300 Å |
Surface Area | 100 m²/g |
Product Code | Brand | Length (Metric) | Diameter (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Diameter (Metric) | Price | Quantity & Availability | ||||||
10344423
|
Thermo Scientific™
73205-052130 |
50 mm | 2.1 mm |
£698.48
Each |
Estimated Shipment: 08-05-2025
Log in to see stock. |
||||||
10150943
|
Thermo Scientific™
73205-053030 |
50 mm | 3 mm |
£738.68
Each |
Estimated Shipment: 08-05-2025
Log in to see stock. |
||||||
10323105
|
Thermo Scientific™
73205-101030 |
100 mm | 1 mm |
£844.20
Each |
Estimated Shipment: 08-05-2025
Log in to see stock. |
||||||
10562455
|
Thermo Scientific™
73205-102130 |
100 mm | 2.1 mm |
£728.63
Each |
Estimated Shipment: 15-05-2025
Log in to see stock. |
||||||
10271275
|
Thermo Scientific™
73205-103030 |
100 mm | 3 mm | N/A | N/A | N/A | |||||
10354133
|
Thermo Scientific™
73205-104630 |
100 mm | 4.6 mm |
£693.45
Each |
Estimated Shipment: 09-05-2025
Log in to see stock. |
||||||
10538205
|
Thermo Scientific™
73205-151030 |
150 mm | 1 mm |
£929.63
Each |
Estimated Shipment: 08-07-2025
Log in to see stock. |
||||||
10602294
|
Thermo Scientific™
73205-152130 |
150 mm | 2.1 mm |
£768.83
Each |
Estimated Shipment: 14-05-2025
Log in to see stock. |
||||||
10699993
|
Thermo Scientific™
73205-153030 |
150 mm | 3 mm | N/A | N/A | N/A | |||||
10181123
|
Thermo Scientific™
73205-154630 |
150 mm | 4.6 mm |
£728.63
Each |
Estimated Shipment: 15-05-2025
Log in to see stock. |
||||||
10496654
|
Thermo Scientific™
73205-251030 |
250 mm | 1 mm |
£974.85
Each |
Estimated Shipment: 29-05-2025
Log in to see stock. |
||||||
10612294
|
Thermo Scientific™
73205-252130 |
250 mm | 2.1 mm | N/A | N/A | N/A | |||||
10572265
|
Thermo Scientific™
73205-254630 |
250 mm | 4.6 mm |
£768.83
Each |
In Stock
Log in to see stock. |
||||||
10725564
|
Thermo Scientific™
73205-054630 |
50 mm | 4.6 mm |
£668.33
Each |
Estimated Shipment: 08-05-2025
Log in to see stock. |
||||||
10331115
|
Thermo Scientific™
73205-011001 |
10 mm | 1 mm | N/A | N/A | N/A | |||||
11538340
|
Thermo Scientific™
73205-014001 |
10 mm | 4 mm |
£383.91
Pack of 4 |
Estimated Shipment: 08-05-2025
Log in to see stock. |
||||||
10545625
|
Thermo Scientific™
73205-013001 |
10 mm | 3 mm | N/A | N/A | N/A | |||||
11528340
|
Thermo Scientific™
73205-012101 |
10 mm | 2.1 mm | N/A | N/A | N/A | |||||
Description
Use Thermo Scientific™ BioBasic™ SCX LC Columns for the optimized separation of proteins, peptides and other cationic species. These high-performance columns are ideal for the analysis of proteins, peptides and other biomolecules with higher efficiency than polymeric ion exchange.
- Strong cation exchange phase based on sulfonic acid chemistry
- Separation and retention of basic and other cationic species
- 300Å pore size for enhanced protein and peptide separations
- Outstanding stability under demanding pH conditions
Ordering Information:
Other column dimensions are available. Please call Customer Service for more information.
Specifications
5800 psi (400 bar) | |
2 to 8 | |
100 m²/g | |
L52 | |
BioBasic | |
Ion Exchange | |
Ion Exchange |
5 μm | |
300 Å | |
60°C | |
Sulfonic Acid | |
3% | |
No |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.