missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ BioBasic™ PREP SEC 300 Size Exclusion Chromatography HPLC Columns

Outstanding size separation of water soluble compounds.

Brand:  Thermo Scientific™ 73505-037821A

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 16671674

  • £571.07 / Each
Estimated Shipment: 23-08-2024
to see stock.
Add to basket

Description

Description

Provide outstanding size separation of water soluble compounds with Thermo Scientific™ BioBasic™ SEC 300 Size Exclusion Chromatography preparative HPLC Columns. BioBasic SEC columns provide high efficiency separations for a wide range of molecular weights from 7,000 to 1,200,000 daltons, allowing for the analysis of proteins, peptides and other biomolecules.

BioBasic™ PREP SEC 300 HPLC Column Benefits:

  • Covers separation of analytes over a wide molecular weight range
  • Long column life and high column efficiencies
  • Simple mechanism of interaction based on molecular size and shape
  • Excellent for sample cleanup
  • Straightforward method development, simple mobile phases
  • 300Å pore size

Ordering Information:

For information on bulk quantities, please inquire.

Specifications

Specifications

Preparative Guard Column
Preparative Size Exclusion
4300 psi (295 bar)
2 to 8
100 m²/g
L59
Spherical, Fully Porous, Ultrapure Silica
Size Exclusion
Size Exclusion
7.8 mm
30 mm
5 μm
300 Å
60°C
Diol (OH)
>2%
No
Preparative Chromatography
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.