Learn More
Thermo Scientific™ BioBasic™ PREP SEC 300 Size Exclusion Chromatography HPLC Columns
Outstanding size separation of water soluble compounds.
£503.92 - £2172.55
Specifications
Particle Size | 5 μm |
---|---|
Pore Size | 300 Å |
USP Type | L59 |
Stationary Phase | Diol (OH) |
Column Type | Size Exclusion |
Product Code | Brand | Particle Size | Diameter (Metric) | Length (Metric) | Column Format | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Particle Size | Diameter (Metric) | Length (Metric) | Column Format | Price | Quantity & Availability | ||
16536130
|
Thermo Scientific™
73505-307846A |
5 μm | 7.8 mm | 300 mm | Preparative Column, Standard |
£2172.55
Each |
Estimated Shipment: 29-07-2024
Log in to see stock. |
||
16681674
|
Thermo Scientific™
73505-157846A |
5 μm | 7.8 mm | 150 mm | Preparative Column, Standard |
£1319.05
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||
16671674
|
Thermo Scientific™
73505-037821A |
5 μm | 7.8 mm | 30 mm | Preparative Guard Column |
£571.07
Each |
Estimated Shipment: 23-08-2024
Log in to see stock. |
||
10301365
|
Thermo Scientific™
73505-037821 |
5 μm | 7.8 mm | 30 mm | Guard Column |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
10119134
|
Thermo Scientific™
73505-157846 |
5 μm | 7.8 mm | 150 mm | High Temperature Column |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
10434413
|
Thermo Scientific™
73505-307846 |
5 μm | 7.8 mm | 300 mm | High Temperature Column |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
16328616
|
Thermo Scientific™
73505-059270A |
5 μm | 20 mm | 50 mm | Preparative Column, Standard |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
Description
Provide outstanding size separation of water soluble compounds with Thermo Scientific™ BioBasic™ SEC 300 Size Exclusion Chromatography preparative HPLC Columns. BioBasic SEC columns provide high efficiency separations for a wide range of molecular weights from 7,000 to 1,200,000 daltons, allowing for the analysis of proteins, peptides and other biomolecules.
BioBasic™ PREP SEC 300 HPLC Column Benefits:
- Covers separation of analytes over a wide molecular weight range
- Long column life and high column efficiencies
- Simple mechanism of interaction based on molecular size and shape
- Excellent for sample cleanup
- Straightforward method development, simple mobile phases
- 300Å pore size
Ordering Information:
For information on bulk quantities, please inquire.
Specifications
5 μm | |
L59 | |
Size Exclusion |
300 Å | |
Diol (OH) | |
Size Exclusion |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.