Learn More
Ansell BioClean-D Long-Length Non-Sterile Hood Product Code.: 16663622
Estimated Shipment: 14-08-2025
to see stock.
Add to basket
Ansell BioClean-D Long-Length Non-Sterile Hood
BIOPHARMA
Product Code. 16663622
Universal size
Click to view available options
Universal size
Product Code. 16663622
Brand: Ansell BDHDL
Estimated Shipment: 14-08-2025
to see stock.
Add to basket
Add to basket
Non-sterile hood features an extra-long yoke for maximum coverage.
  • Lightweight low-linting CleanTough material
  • ESD properties
  • PPE Cat 3 Type PB [6]
  • Silicone free
  • Certifications and Testing:
    • Manufactured in a facility holding ISO 9001:2015 and (EU)2016/425
    • Module D certificationsConforms to Category 3 - Personal Protective Equipment Regulation(EU)2016/425
    • Tested against standard EN ISO 13034:2005+A1:2009 protective clothing against liquid chemicals (Type PB[6])
    • Tested against standard EN ISO 14325 protective clothing against chemicals
    • Tested against standards: EN 1149-5:2008, protective clothing -electrostatic properties; EN 530, Abrasion resistance; EN 7854, Flexcracking resistance; EN 6530, Resistance to penetration by liquids; EN13934-1, Tensile strength; EN 9073-4, Trapezoidal tear resistance; EN863, Puncture resistance; and EN 13935-2, Seam strength.
    • Tested against permeation standard ASTM F739-12
    • Processed in an NEBB certified ISO Class 4 environment
TRUSTED_SUSTAINABILITY
Specifications
Product Type Hood
Size Universal
Material Spunbond Polypropylene
Color White
Closure Type Elastic
Disposable Single-use
Design Type Three-piece
Model BDHD
Seam Type Bound, Single Needle Stitching
Sterility Non-sterile
Quantity 120/Pk.
Hood Type Three-piece, Elastic face-opening
Show More Show Less
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number25-0434 Certificate TypeCertificate of Compliance Date01/04/2025 Catalog NumberBDHD-L
Lot Number24-3335 Certificate TypeCertificate of Compliance Date04/03/2025 Catalog NumberBDHD-L
Lot Number24-2069 Certificate TypeCertificate of Compliance Date18/02/2025 Catalog NumberBDHD-L
Lot Number24-1706 Certificate TypeCertificate of Compliance Date18/02/2025 Catalog NumberBDHD-L
Lot Number24-1206 Certificate TypeCertificate of Compliance Date18/02/2025 Catalog NumberBDHD-L
Product Content Correction
Product Title
Ansell BioClean-D Long-Length Non-Sterile Hood >