Learn More
Corning™ BioCoat™ Collagen IV TC-Treated Culture Dishes
Designed to closely mimic in vivo environments and enhance expression of cell-specific morphology
Brand:  Corning™ 354416
Code : 67
Additional Details : Weight : 3.06170kg
 View more versions of this product
Product Code. 10142861
  • £213.52 / Pack of 20
Estimated Shipment: 26-11-2024
to see stock.
Add to basket
Description
Description
Microporous membranes permit diffusion of ions, low-molecular-weight proteins and other substances to both apical and basolateral cell surfaces, simulating the natural environment of cells in vivo.
  • Source: Mouse
  • TC-treated
  • Nonsterile
For adhesion, growth and differentiation of neuronal and transfected cell lines
Specifications
Specifications
Round
Polystyrene
Collagen Type IV
20/Cs.
60 mm
21.3 cm2
Round
Cell Culture Dish
5/Pk.