missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ BioCoat™ Collagen IV TC-Treated Culture Dishes
Designed to closely mimic in vivo environments and enhance expression of cell-specific morphology
£199.02 - £213.52
Specifications
Bottom Shape | Round |
---|---|
Material | Polystyrene |
Shape | Round |
Surface Treatment | Collagen Type IV |
Type | Cell Culture Dish |
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
10142861
|
Corning™
354416 |
60 mm |
£213.52
Pack of 20 |
Estimated Shipment: 26-11-2024
Log in to see stock. |
|||||
10479392
|
Corning™
354459 |
35 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Microporous membranes permit diffusion of ions, low-molecular-weight proteins and other substances to both apical and basolateral cell surfaces, simulating the natural environment of cells in vivo.
- Source: Mouse
- TC-treated
- Nonsterile
For adhesion, growth and differentiation of neuronal and transfected cell lines
Specifications
Round | |
Round | |
Cell Culture Dish |
Polystyrene | |
Collagen Type IV |