missing translation for 'onlineSavingsMsg'
Learn More

Corning™ BioCoat™ Poly-D-Lysine Vented Cap Flasks

Ideal for applications involving adhesion, growth and differentiation of neuronal and transfected cell lines

Brand:  Corning™ 354539

Code : 66

Additional Details : Weight : 0.95000kg

 View more versions of this product

Product Code. 10626691

  • £116.99 / Pack of 5
Estimated Shipment: 26-11-2024
to see stock.
Add to basket

Description

Description

Corning™ BioCoat™ products come in a variety of pre-coated options and are an ideal solution for enhanced cell attachment and growth of a variety of primary cells and transformed cells in serum-free or serum-containing cultures.
Specifications

Specifications

25.3 oz.
750 mL
Canted
Rectangular
Sterile
Phenolic
For adhesion, growth and differentiation of neuronal and transfected cell lines
5/cs.
Poly-D-Lysine
175 cm2
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Disclaimer: This product is not intended to mitigate the presence of microorganisms on surfaces or in the environment, where such organisms can be deleterious to humans or the environment.