missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ BioCoat™ Poly-D-Lysine Vented Cap Flasks
Ideal for applications involving adhesion, growth and differentiation of neuronal and transfected cell lines
£80.02 - £695.57
Specifications
Closure Material | Phenolic |
---|---|
For Use With (Application) | For adhesion, growth and differentiation of neuronal and transfected cell lines |
Neck Style | Canted |
Shape | Rectangular |
Surface Coating | Poly-D-Lysine |
Product Code | Brand | Culture Area | No. per Case | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Culture Area | No. per Case | Price | Quantity & Availability | ||||
10666111
|
Corning™
354537 |
75 cm2 | 5/cs. |
£80.02
Pack of 5 |
Estimated Shipment: 26-11-2024
Log in to see stock. |
||||
10297280
|
Corning™
354538 |
150 cm2 | 5/cs. |
£104.62
Pack of 5 |
Estimated Shipment: 26-11-2024
Log in to see stock. |
||||
10626691
|
Corning™
354539 |
175 cm2 | 5/cs. |
£116.99
Pack of 5 |
Estimated Shipment: 26-11-2024
Log in to see stock. |
||||
10796672
|
Corning™
356537 |
75 cm2 | 50/cs. |
£510.92
Pack of 50 |
Estimated Shipment: 26-11-2024
Log in to see stock. |
||||
10410102
|
Corning™
356539 |
175 cm2 | 40/cs. |
£695.57
Pack of 40 |
Estimated Shipment: 26-11-2024
Log in to see stock. |
||||
10397941
|
Corning™
356536 |
25 cm2 | 50/cs. |
£449.40
Pack of 50 |
Estimated Shipment: 26-11-2024
Log in to see stock. |
||||
10400102
|
Corning™
356538 |
150 cm2 | 40/cs. |
£652.47
Pack of 40 |
Estimated Shipment: 26-11-2024
Log in to see stock. |
||||
10711371
|
Corning™
354536 |
25 cm2 | 10/cs. |
£116.99
Pack of 10 |
Estimated Shipment: 26-11-2024
Log in to see stock. |
||||
Description
Corning™ BioCoat™ products come in a variety of pre-coated options and are an ideal solution for enhanced cell attachment and growth of a variety of primary cells and transformed cells in serum-free or serum-containing cultures.Specifications
Phenolic | |
Canted | |
Poly-D-Lysine |
For adhesion, growth and differentiation of neuronal and transfected cell lines | |
Rectangular | |
Sterile |
Disclaimer: This product is not intended to mitigate the presence of microorganisms on surfaces or in the environment, where such organisms can be deleterious to humans or the environment.