Learn More
Fisherbrand™ Biohazard Labels
Bright red fluorescent labels display the universal biohazard symbol for easy identification.
Brand: Fisherbrand™
Additional Details : Weight : 1.00000kg
Description
Bright red fluorescent labels display the universal biohazard symbol for easy identification.
- Hazard communication labels ensure safety and efficiency
- Identify equipment, containers, rooms, materials, experimental animals or combinations of these items that may contain potentially infectious agents
- Easy peel-off labels with pressure-sensitive adhesive
Use in accordance with applicable federal and state regulations.
Specifications
Pressure-sensitive | |
Label | |
Biohazard Warning Tape | |
500/Pk. | |
3 in. | |
2 in. |
Fluorescent Red, Black | |
Paper | |
Biohazard | |
3 in. x 2 in. | |
7.62 cm |