missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Biohazard Labels
Bright red fluorescent labels display the universal biohazard symbol for easy identification.
£10.46
Specifications
Adhesive Type | Pressure-sensitive |
---|---|
Format | Label |
Material | Paper |
Product Type | Biohazard Warning Tape |
For Use With (Application) | Biohazard |
Product Code | Brand | Quantity | Length (English) | Length (Metric) | Width (English) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Length (English) | Length (Metric) | Width (English) | Price | Quantity & Availability | ||
17825838
|
Fisherbrand™
|
500/Pk. | 3 in. | 7.62 cm | 2 in. |
£10.46
Pack of 500 |
Estimated Shipment: 01-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
Description
Bright red fluorescent labels display the universal biohazard symbol for easy identification.
- Hazard communication labels ensure safety and efficiency
- Identify equipment, containers, rooms, materials, experimental animals or combinations of these items that may contain potentially infectious agents
- Easy peel-off labels with pressure-sensitive adhesive
Use in accordance with applicable federal and state regulations.
Specifications
Pressure-sensitive | |
Paper | |
Biohazard |
Label | |
Biohazard Warning Tape |