Learn More
Thermo Scientific™ BIOLiner™ Rotor Bucket Adapters
Combine these rotor bucket adapters with the BIOLiner Rotor Bucket for a complete system.
Brand: Thermo Scientific™ 75003671
453.14 GBP valid until 2025-07-13
Use promo code "24135" to get your promotional price.
Description
Combine Thermo Scientific™ BIOLiner™ Rotor Bucket Adapters with the BIOLiner Rotor Bucket for a complete system.
Ordering Information:
Individual adapters and bottle/adapter packages are offered.
- Capacity range: 3L to 980mL
- Maximum tube dimensions (O.D. x L): 98 × 124mm
Includes:
75-003-692 includes 750mL flat-bottom bottle with all round adapters for the TX-750 round bucket and four 500mL Corning™ conical bottle adapters.

Specifications
Round Bucket Adapter | |
5/7 mL Blood Collection Tube | |
110 mm |
5/7 mL Blood Collection Tube | |
14 mm | |
4 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.