Learn More
Sartorius™ Biosart™ Monitor
Microbiological and analytical filtration units offering superior performance
Brand: Sartorius™ 16403-47-04----K
171.99 GBP valid until 2025-07-13
Use promo code "24135" to get your promotional price.
This item is not returnable.
View return policy
Description
The system enables the user to select the optimal hardware and consumables for their needs in quality assurance. At the heart of the system is a stainless steel manifold designed to accommodate all types of filter holder and funnels. Please note: grey filters turn black when wet. Multi-branch system for routine colony or particle counting.
Specifications
Gray/White | |
48Pack | |
2 layers of 24 | |
Monitor |
Sterile | |
48Pack | |
48Pack |
Product Content Correction
Product Title
Sartorius™ Biosart™ Monitor > Color: Grey/White
Spot an opportunity for improvement?Share a Content Correction