Learn More
Thermo Scientific™ BIOShield™ 720 Rotor Tube Adapters
Enhance your rotor's versatility with Thermo Scientific™ BIOShield™ 720 Tube Adapters, which support a wide assortment of tube sizes.
£446.68 - £517.03
Products 13
Product Code Brand Description Price Quantity & Availability  
Product Code Brand Description Price Quantity & Availability  
10247154
View Documents
Thermo Scientific™
75003677
50mL Conical
£468.72
Pack of 4
10597004
View Documents
Thermo Scientific™
75003678
15mL Conical
£467.87
Pack of 4
10657555
View Documents
Thermo Scientific™
75003693
180mL Polypropylene Screw Cap (Bottle)
£501.78
Pack of 4
10124354
View Documents
Thermo Scientific™
75003701
10mL Blood Collection
£448.38
Pack of 4
10083114
View Documents
Thermo Scientific™
75003813
150mL Round-bottom, Open-tube This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10546794
View Documents
Thermo Scientific™
75003814
100mL Round-bottom, Open-tube
£455.15
Pack of 4
10625845
View Documents
Thermo Scientific™
75003816
50mL DIN Round-bottom
£468.72
Pack of 4
10757395
View Documents
Thermo Scientific™
75003817
25mL DIN Round-bottom
£446.68
Pack of 4
10757585
View Documents
Thermo Scientific™
75003818
25mL Universal
£517.03
Pack of 4
10555284
View Documents
Thermo Scientific™
75003820
15mL Starstedt
£472.11
Pack of 4
10104754
View Documents
Thermo Scientific™
75003821
5/7mL Blood Collection
£446.68
Pack of 4
10546434
View Documents
Thermo Scientific™
75003822
5/7mL Round-bottom, Open-tube
£467.87
Pack of 4
10154464
View Documents
Thermo Scientific™
75003823
1.5/2.0mL Conical Microtube
£455.15
Pack of 4
Description
Description
Enhance your rotor's versatility with Thermo Scientific™ BIOShield™ 720 Tube Adapters, which support a wide assortment of tube sizes. These adapters are for use with the BIOShield 720 High Speed Swinging-Bucket Rotor.
  • Maximum tube capacity: 4 × 150mL
  • Maximum O.D. × L: 50 × 122mm