Learn More
Agilent BioTek Complete Waste System
Provides supply and waste containment with optimal evacuation efficiency for 384- and 1536-well microplates
Brand: Agilent BioTek 7100752
This item is not returnable.
View return policy
Description
- Includes complete high flow waste system and complete dispense system
- 115V/230V
Specifications
115V high flow vacuum pump assembly, vacuum set with level sense assembly with 10 L bottles, dispense system complete assembly with 10 L bottles | |
115/230 V | |
Complete Dispense/High Flow Waste System | |
EL406 Microplate Washer, ELx 405 Microplate Washer |