Learn More
Agilent BioTek Complete Waste System
GREENER_CHOICE
Provides supply and waste containment with optimal evacuation efficiency for 384- and 1536-well microplates
£7975.00 - £8697.00
Products 2
Product Code Brand Includes For Use With (Equipment) Type Price Quantity & Availability  
Product Code Brand Includes For Use With (Equipment) Type Price Quantity & Availability  
15645437
GREENER_CHOICE
View Documents
Agilent BioTek
7100751
ELx 405 Microplate Washer Complete Dispense/High Flow Waste System
£7975.00
Each
16522252
GREENER_CHOICE
View Documents
Agilent BioTek
7100752
115V high flow vacuum pump assembly, vacuum set with level sense assembly with 10 L bottles, dispense system complete assembly with 10 L bottles EL406 Microplate Washer, ELx 405 Microplate Washer Complete Dispense/High Flow Waste System
£8697.00
Each
Description
Description
  • Includes complete high flow waste system and complete dispense system
  • 115V/230V