missing translation for 'onlineSavingsMsg'
Learn More

IBA Lifesciences Biotin stock solution
GREENER_CHOICE

Product Code. 17207600 Shop All IBA Lifesciences Products
Click to view available options
:
100 mM
This item is not returnable. View return policy

Product Code. 17207600

Brand: IBA Lifesciences 66325001

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For dissociation of Twin-Strep-tag™ from Strep-Tactin™ suitable for cells

  • Biotin stock solution for preparing a dissociation buffer when using the Strep-tag™ technology for cells (Fab-TACS™/Nano-TACS™, CD3/CD28 Streptamer™, MHC I Streptamer™).
  • Biotin addition causes the rapid dissociation of isolation reagents from the cell surface.
  • Thus, fully functional, highly pure, and non-induced cells are received

Specifications

Concentration 100 mM
Formulation Dissolved in DPBS

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.