missing translation for 'onlineSavingsMsg'
Learn More
Bis(2,4,6-trichlorophenyl)oxalate, 98%, Thermo Scientific Chemicals Product Code.: 10085760

Bis(2,4,6-trichlorophenyl)oxalate, 98%, Thermo Scientific Chemicals

Product Code. 10085760
5 g, Glass bottle
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10085760

Brand: Thermo Scientific Acros 402660050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1165-91-9
CAS Min % 97.5
CAS Max % 100.0
Melting Point 188°C to 192°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C14H4Cl6O4
MDL Number MFCD00043061
Quantity 5 g
Synonym bis 2,4,6-trichlorophenyl oxalate, bis 2,4,6-trichlorophenyl ethanedioate, oxalic acid bis 2,4,6-trichlorophenyl ester, 2,4,6-trichlorophenyl oxalate, ethanedioic acid, bis 2,4,6-trichlorophenyl ester, bis-tcpo, acmc-20a7jt, gevpiwpywjzspr-uhfffaoysa
InChI Key GEVPIWPYWJZSPR-UHFFFAOYSA-N
SMILES C1=C(C=C(C(=C1Cl)OC(=O)C(=O)OC2=C(C=C(C=C2Cl)Cl)Cl)Cl)Cl
IUPAC Name bis(2,4,6-trichlorophenyl) oxalate
Molecular Weight (g/mol) 448.89
PubChem CID 160567
Formula Weight 448.89
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Bis(2, 4, 6-trichlorophenyl)oxalate, 98%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0468092 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number402660050
Lot NumberA0454701 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number402660050
Lot NumberA0396221 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number402660050
Lot NumberA0411185 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number402660050
Lot NumberA0405055 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number402660050
compliance-icons
Product Identifier
  • Bis(2,4,6-trichlorophenyl)oxalate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Bis(2,4,6-trichlorophenyl)oxalate, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.