Learn More
Bis(methylthio)methane, 99%, Thermo Scientific Chemicals
£20.80 - £45.10
Chemical Identifiers
CAS | 1618-26-4 |
---|---|
Molecular Formula | C3H8S2 |
Molecular Weight (g/mol) | 108.217 |
MDL Number | MFCD00008564 |
InChI Key | LOCDPORVFVOGCR-UHFFFAOYSA-N |
Synonym | bis methylthio methane, bis methylsulfanyl methane, 2,4-dithiapentane, methane, bis methylthio, bis methylmercapto methane, formaldehyde dimethyl mercaptal, methylenebis methyl sulfide, thioformaldehyde dimethylacetal, ch3sch2sch3 |
PubChem CID | 15380 |
IUPAC Name | bis(methylsulfanyl)methane |
SMILES | CSCSC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11448597
|
Thermo Scientific Alfa Aesar
A19706.06 |
5 g |
£20.80
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11458597
|
Thermo Scientific Alfa Aesar
A19706.14 |
25 g |
£45.10
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1618-26-4 | |
108.217 | |
LOCDPORVFVOGCR-UHFFFAOYSA-N | |
15380 | |
CSCSC |
C3H8S2 | |
MFCD00008564 | |
bis methylthio methane, bis methylsulfanyl methane, 2,4-dithiapentane, methane, bis methylthio, bis methylmercapto methane, formaldehyde dimethyl mercaptal, methylenebis methyl sulfide, thioformaldehyde dimethylacetal, ch3sch2sch3 | |
bis(methylsulfanyl)methane |
Specifications
1618-26-4 | |
43°C (109°F) | |
C3H8S2 | |
UN1993 | |
14,1256 | |
LOCDPORVFVOGCR-UHFFFAOYSA-N | |
bis(methylsulfanyl)methane | |
15380 | |
99% |
146°C to 147°C | |
Unpleasant | |
MFCD00008564 | |
1731143 | |
bis methylthio methane, bis methylsulfanyl methane, 2,4-dithiapentane, methane, bis methylthio, bis methylmercapto methane, formaldehyde dimethyl mercaptal, methylenebis methyl sulfide, thioformaldehyde dimethylacetal, ch3sch2sch3 | |
CSCSC | |
108.217 | |
108.23 | |
Bis(methylthio)methane |
Safety and Handling
GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
missing translation for 'tsca' : TSCA
RUO – Research Use Only