Learn More
Bis(triphenylphosphine)dicarbonylnickel, 98%, Thermo Scientific Chemicals
£34.20 - £299.00
Chemical Identifiers
CAS | 13007-90-4 |
---|---|
Molecular Formula | C38H30NiO2P2 |
Molecular Weight (g/mol) | 639.30 |
MDL Number | MFCD00009591 |
InChI Key | SLFKPACCQUVAPG-UHFFFAOYSA-N |
Synonym | bis triphenylphosphine dicarbonylnickel |
PubChem CID | 22836333 |
IUPAC Name | carbon monoxide;nickel;triphenylphosphanium |
SMILES | [Ni].[C-]#[O+].[C-]#[O+].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11323638
|
Thermo Scientific Alfa Aesar
039242.27 |
2.5 g |
£34.20
2.50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11313638
|
Thermo Scientific Alfa Aesar
039242.09 |
10 g |
£83.20
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11333638
|
Thermo Scientific Alfa Aesar
039242.18 |
50 g |
£299.00
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13007-90-4 | |
639.30 | |
SLFKPACCQUVAPG-UHFFFAOYSA-N | |
22836333 | |
[Ni].[C-]#[O+].[C-]#[O+].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1 |
C38H30NiO2P2 | |
MFCD00009591 | |
bis triphenylphosphine dicarbonylnickel | |
carbon monoxide;nickel;triphenylphosphanium |
Specifications
13007-90-4 | |
C38H30NiO2P2 | |
UN3466 | |
bis triphenylphosphine dicarbonylnickel | |
[Ni].[C-]#[O+].[C-]#[O+].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1 | |
639.30 | |
639.29 | |
Crystalline |
210°C to 215°C (decomposition) | |
MFCD00009591 | |
14,1304 | |
SLFKPACCQUVAPG-UHFFFAOYSA-N | |
carbon monoxide;nickel;triphenylphosphanium | |
22836333 | |
98% | |
Bis(triphenylphosphine)dicarbonylnickel |
Safety and Handling
GHS H Statement
H300-H310-H330
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
P201-P202-P261-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
H317-H350
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: METAL CARBONYLS, SOLID, N.O.S.
missing translation for 'einecsNumber' : 235-851-9
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only