missing translation for 'onlineSavingsMsg'
Learn More
Bismuth pieces, 10cm (3.9in) & down, 99.99% (metals basis), Thermo Scientific Chemicals Product Code.: 11306618

Bismuth pieces, 10cm (3.9in) & down, 99.99% (metals basis), Thermo Scientific Chemicals

Product Code. 11306618
1 kg
Click to view available options
Quantity:
250 g
1 kg
5 kg
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 11306618

Brand: Thermo Scientific Alfa Aesar 012208.A1

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-69-9
Molecular Formula Bi
Molecular Weight (g/mol) 208.98
MDL Number MFCD00134033
InChI Key JCXGWMGPZLAOME-UHFFFAOYSA-N
Synonym bismuth, elemental, unii-u015tt5i8h, powder, iii ion, shot, elongated, rod, 12.7mm 0.5in dia, atom, needles, pieces, rod
PubChem CID 5359367
ChEBI CHEBI:33301
IUPAC Name bismuth
SMILES [Bi]

Specifications

Color Gray
Appearance Gray needles
Assay Percent Range 99.99% (Metals basis)
Quantity 1 kg
Odor Odorless
Vapor Pressure 1 hPa (1mm Hg)
Form Pieces
Chemical Name or Material Bismuth

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberM07L009 Certificate TypeCertificate of Analysis Date09/01/2025 Catalog Number012208.A1,012208.30,012208.A7
Lot NumberZ02K043 Certificate TypeCertificate of Analysis Date11/12/2024 Catalog Number012208.A1,012208.30,012208.A7
Lot NumberY19H048 Certificate TypeCertificate of Analysis Date23/08/2022 Catalog Number012208.A1,012208.30,012208.A7,012216.GH
Lot NumberN21I055 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number012208.A1,012186.18,012186.09,012186.30
Lot NumberH03Q09 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number012208.A1,012208.30,012208.A7,012216.GH

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Bismuth pieces, 10cm (3.9in) & down, 99.99% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.