Learn More
Thermo Scientific™ Black 96-Well Immuno Plates

Perform quantitative and qualitative immunoassays, like ELISA and binding assays, with these black, 96-well immuno plates, available in a choice of three surface types.
Brand: Thermo Scientific™ 7805
174.00 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Perform quantitative and qualitative immunoassays, like ELISA and binding assays, with Thermo Scientific™ Black 96-Well Immuno Plates. Featuring a choice of three surface types and optimal binding, these plates minimize background in fluorescence reading.
Features:
- Plate has SBS footprint — fits standard equipment
- Flat-bottom wells
- Certified binding homogeneity/reproducibility
Choice of surface:
- Thermo Scientific PolySorp, Microfluor 1, Microlite 1+ and Universal Binding (UB) are for absorption of hydrophobic molecules.
- Thermo Scientific MaxiSorp is hydrophilic and ideal for antibody sandwich assays.
- Microfluor 2 and Microlite 2+ are slightly hydrophilic and bind a diverse range of biomolecules.
Compatible with:
Compatible with lids for Thermo Scientific MicroWell plates, sealing tape and breathable membrane
Ordering Information:
Availability may vary by country

Specifications
Slightly Hydrophilic | |
Without Lid | |
8 x 12 | |
Black | |
Polystyrene | |
96 | |
High Binding | |
Fluorescence | |
10 | |
Antibodies (Glycoproteins) and Proteins |
Standard | |
Fluorescence | |
50 Cases | |
Microplate | |
330 μL | |
280 μL | |
Flat Bottom (F-well) | |
50 | |
None | |
F96 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.