Learn More
Thermo Scientific™ Black 96-Well Immuno Plates

Perform quantitative and qualitative immunoassays, like ELISA and binding assays, with these black, 96-well immuno plates, available in a choice of three surface types.
Brand: Thermo Scientific™ 9502867
140.99 GBP valid until 2025-03-28
Use promo code "24107" to get your promotional price.
Description
Perform quantitative and qualitative immunoassays, like ELISA and binding assays, with Thermo Scientific™ Black 96-Well Immuno Plates. Featuring a choice of three surface types and optimal binding, these plates minimize background in fluorescence reading.
Features:
- Plate has SBS footprint — fits standard equipment
- Flat-bottom wells
- Certified binding homogeneity/reproducibility
Choice of surface:
- Thermo Scientific PolySorp, Microfluor 1, Microlite 1+ and Universal Binding (UB) are for absorption of hydrophobic molecules.
- Thermo Scientific MaxiSorp is hydrophilic and ideal for antibody sandwich assays.
- Microfluor 2 and Microlite 2+ are slightly hydrophilic and bind a diverse range of biomolecules.
Compatible with:
Compatible with lids for Thermo Scientific MicroWell plates, sealing tape and breathable membrane
Ordering Information:
Availability may vary by country

Specifications
Hydrophobic | |
Without Lid | |
8 x 12 | |
Adsorption | |
Microplate | |
350 μL | |
350 μL | |
Universal Binding | |
Fluorescence | |
25 | |
50/Cs. |
Standard | |
Fluorescence | |
50 Cases | |
Black | |
Polystyrene | |
96 | |
Non-sterile | |
Flat Bottom (F-well) | |
50 | |
None | |
Lipids |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.