missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hellma Black Aluminum Spacer for Spectrophotometer
Suitable for fitting a smaller sized optical path cell into a 10 mm cell holder.
Brand: Hellma 013-102-71
This item is not returnable.
View return policy
Description
- Aluminum construction
- Suitable for any beam height
- Available for 1 mm, 2 mm, or 5 mm path length cells/cuvettes
Specifications
38 x 12.5 x 9 mm | |
2 mm cell | |
2 mm |
10 mm cell holder, spectrophotometer | |
Aluminum |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction