missing translation for 'onlineSavingsMsg'
Learn More

Hellma Black Aluminum Spacer for Spectrophotometer

Suitable for fitting a smaller sized optical path cell into a 10 mm cell holder.

Brand:  Hellma 013-105-71

 View more versions of this product

Product Code. 15718032

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

  • Aluminum construction
  • Suitable for any beam height
  • Available for 1 mm, 2 mm, or 5 mm path length cells/cuvettes
Specifications

Specifications

38 x 12.5 x 9 mm
5 mm cell
5 mm
10 mm cell holder, spectrophotometer
Aluminum
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.