missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ BLAUBRAND™ Graduated Measuring Pipettes

AR-GLAS™ construction. BLAUBRAND™ Graduated Measuring Pipettes are calibrated to contain (TC, In.)

£96.95 - £120.00

Specifications

Calibrated Yes
Calibration To Contain
Class A
Material AR-GLAS
Graduated Yes
View More Specs

Products 2
Product Code Brand Volume (Metric) Price Quantity & Availability  
Product Code Brand Volume (Metric) Price Quantity & Availability  
11835830
View Documents Promotion Details
BRAND™
27704
0.2 mL
£96.95
Pack of 12
In Stock
to see stock.
Add to basket
 
11818311
View Documents Promotion Details
BRAND™
27702
0.1 mL
£120.00
Pack of 12
Estimated Shipment: 29-05-2025
to see stock.
Add to basket
 
Description

Description

  • AR-GLAS™ construction
  • Class A, DE-M marking
  • Error limits and sub-division according to DIN 12689
  • Calibrated to contain (TC, In)
  • All BLAUBRAND™ graduated pipettes are delivered with batch number and enclosed one batch certificate per packing unit of the manufacturer.
  • On demand also available with individual certificate, USP Individual certificate or DAkkS calibration certificate.
Specifications

Specifications

Yes
A
Yes
12/Pk.
To Contain
AR-GLAS
Blue
360 mm
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
BRAND™ BLAUBRAND™ Graduated Measuring Pipettes

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.