Learn More
BRAND™ BLAUBRAND™ Class A Borosilicate Glass Mixing Cylinders
Includes polypropylene stopper and hexagonal base. BRAND™ Blaubrand™ Class A Borosilicate Glass Mixing Cylinders are calibrated to contain the liquid inside the vessel.
Brand:  BRAND™ 32408
39.61 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 11828341
  • £62.90 / Pack of 2
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Borosilicate glass construction (Boro 3.3. DIN EN ISO 4788)
  • Calibrated to contain (TC, In)
  • Includes polypropylene stopper and hexagonal base
  • Height without stopper
  • All BLAUBRAND™ cylinders are delivered with batch number and enclosed one batch certificate per packing unit of the manufacturer
  • On demand also available with individual certificate, USP Individual certificate or DAkkS calibration certificate
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Hexagonal
Blue
Graduated Mixing Cylinder
DIN EN ISO 4788
10 mL
Batch Certificate, Polypropylene Stopper
Polypropylene
0.2 mL
To Contain
Round
Borosilicate Glass
B
160 mm
Blue Enamel Graduations
Polypropylene
Autoclavable