missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Blaubrand™ ETERNA™ Class A USP Volumetric Flasks, with Certificate

Constructed of Borosilicate Glass and complies with DIN EN ISO 1042 standards. Calibrated to contain a precise amount of liquid at a specific temperature. Includes polypropylene stopper and DE-M marking. Delivered with individual conformity certificate.

Brand:  BRAND™ 956847

Additional Details : Weight : 0.09000kg

 View more versions of this product

Product Code. 15875562

  • £45.35 / Each
Estimated Shipment: 20-08-2024
to see stock.
Add to basket

Description

Description

  • Class A Volumetric Flask
  • Constructed of Borosilicate Glass
  • Complies with DIN EN ISO 1042 standards
  • Calibrated to contain (TC, In)
  • Narrow neck and wide neck options available in a variety of sizes
  • Includes polypropylene stopper and DE-M marking
  • Delivered with individual USP conformity certificate
Specifications

Specifications

Calibrated To Contain
Polypropylene
50 mL
±0.06 mL
Yes
Stopper
USP individual and USP batch-certificate, DIN EN ISO 1042
Clear
Borosilicate Glass
Narrow
A
12/21
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers