Learn More
BRAND™ BLAUBRAND™ Glass Graduated Cylinders, Class A, USP, Certified
Class A, USP, certified graduated cylinders
£34.40 - £155.00
Specifications
Class A
Material Borosilicate Glass
View More Specs
Products 3
Product Code Brand Capacity (Metric) Graduations Price Quantity & Availability  
Product Code Brand Capacity (Metric) Graduations Price Quantity & Availability  
15734037
View Documents Promotion Details
BRAND™
32820
25 mL 0.5 mL
£34.40
Pack of 2
In Stock
to see stock.
Add to basket
15358286
View Documents Promotion Details
BRAND™
32854
500 mL 5 mL
£128.00
Pack of 2
In Stock
to see stock.
Add to basket
15584409
View Documents Promotion Details
BRAND™
32864
2000 mL 20 mL
£155.00
Each
In Stock
to see stock.
Add to basket
Description
Description
  • Made of low expansion coefficient, chemically resistant Borosilicate 3.3 glass
  • Available in all ISO sizes from 5 to 2000mL
  • Blue enamel print represents optimum combination on resistance and readability
  • Supplied with batch number and an accompanying USP batch certificate in original packaging
Specifications
Specifications
A
Borosilicate Glass
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
BRAND™ BLAUBRAND™ Glass Graduated Cylinders, Class A, USP, Certified