missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Blocker™ Casein

Blocker Casein is a ready-to-use solution of purified casein for blocking steps in western blot, ELISA, IHC, and nucleic acid detection methods.

Brand:  Thermo Scientific™ 37582

39.23 GBP valid until 2024-12-31
Use promo code "21651" to get your promotional price.


 View more versions of this product

Product Code. 13454299

  • £55.25 / 100mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Blocker Casein is a ready-to-use solution of purified casein for blocking steps in western blot, ELISA, IHC, and nucleic acid detection methods. Blocker Casein is 1% (w/v) casein, which corresponds to the optimal concentration for most applications.

Features of Blocker Casein include:
Purified casein—single-protein blocking buffer provides fewer chances of cross-reaction with assay components than serum or milk solutions
Easy to use—1% casein solution is ready to use; can be diluted further as needed
Safe—stable, thimerosal-free formulation

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Blocker Casein in PBS, 100 mL
100 mL
Western Blot, ELISA
1X
Single purified protein
Blocker Casein
Liquid
Blocker™
PBS
Store at 4°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.