missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Blocker™ Casein
Blocker Casein is a ready-to-use solution of purified casein for blocking steps in western blot, ELISA, IHC, and nucleic acid detection methods.
£53.75 - £178.00
Specifications
Product Type | Blocker Casein |
---|---|
Form | Liquid |
For Use With (Application) | Western Blot, ELISA |
Product Line | Blocker™ |
Concentration | 1X |
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
13454299
|
Thermo Scientific™
37582 |
Blocker Casein in PBS, 100 mL |
£55.25
100mL |
Estimated Shipment: 29-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10496134
|
Thermo Scientific™
37528 |
Blocker Casein in PBS, 1 L |
£178.00
1 L |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13464299
|
Thermo Scientific™
37583 |
Blocker Casein in TBS, 100 mL |
£53.75
100mL |
Estimated Shipment: 05-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10443644
|
Thermo Scientific™
37532 |
Blocker Casein in TBS, 1 L |
£174.00
1 L |
Estimated Shipment: 25-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Blocker Casein is a ready-to-use solution of purified casein for blocking steps in western blot, ELISA, IHC, and nucleic acid detection methods. Blocker Casein is 1% (w/v) casein, which corresponds to the optimal concentration for most applications.
Features of Blocker Casein include:• Purified casein—single-protein blocking buffer provides fewer chances of cross-reaction with assay components than serum or milk solutions
• Easy to use—1% casein solution is ready to use; can be diluted further as needed
• Safe—stable, thimerosal-free formulation
Specifications
Blocker Casein | |
Western Blot, ELISA | |
1X | |
Store at 4°C. |
Liquid | |
Blocker™ | |
Single purified protein |