Learn More
Fisherbrand™ Blocks for Fisherbrand™ Isotemp™ Digital Dry Baths/Block Heaters-Mixed Blocks
Constructed of aluminum alloy
£97.75 - £182.00
Specifications
For Use With (Equipment) | Thermo Scientific Digital and Touch Screen Dry Baths/Block Heaters (No compact models) |
---|---|
Material | Aluminum Alloy |
Type | Block |
Product Code | Brand | No. of Places | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Places | Price | Quantity & Availability | ||||||
15337948
|
Fisherbrand™
88860112 |
32 |
£132.00
Each |
Estimated Shipment: 05-11-2025
Log in to see stock. |
||||||
15347948
|
Fisherbrand™
88860113 |
18 |
£132.00
Each |
Estimated Shipment: 01-07-2025
Log in to see stock. |
||||||
15357948
|
Fisherbrand™
88860114 |
3 |
£133.00
Each |
Estimated Shipment: 30-05-2025
Log in to see stock. |
||||||
15377948
|
Fisherbrand™
88860115 |
30 |
£182.00
Each |
Estimated Shipment: 26-09-2025
Log in to see stock. |
||||||
15327948
|
Fisherbrand™
88860111 |
1 |
£97.75
Each |
Estimated Shipment: 26-09-2025
Log in to see stock. |
||||||
Description
A variety of blocks for any application
Specifications
Thermo Scientific Digital and Touch Screen Dry Baths/Block Heaters (No compact models) | |
Aluminum Alloy | |
Block |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Fisherbrand™ Blocks for Fisherbrand™ Isotemp™ Digital Dry Baths/Block Heaters-Mixed Blocks